accessibility menu, dialog, popup

UserName

Viewing profile as user:
NFFFFFCl

CAS RN 118559-21-0

IUPAC Name: 2-chloro-5-(1,1,2,2,2-pentafluoroethyl)pyridine
Synonyms: 2-chloro-5-(1,1,2,2,2-pentafluoroethyl)pyridine
Molecular Weight (g/mol): 231.55
Molecular Formula: C7H3ClF5N
InChi Key: YRBHRYDSCMEMAY-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C1=CN=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results