accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)OH(S)HNOHOOO

CAS RN 118609-38-4

IUPAC Name: (2S,3R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-hydroxybutanoic acid
Synonyms: (2S,3R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-hydroxybutanoic acid|FMOC-L-THR-OH
Molecular Weight (g/mol): 341.36
Molecular Formula: C19H19NO5
InChi Key: OYULCCKKLJPNPU-DIFFPNOSSA-N
SMILES: C[C@@H](O)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results