accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1186103-43-4

IUPAC Name: benzyl(ethyl)dimethylazanium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms: benzyl(ethyl)dimethylazanium; bistriflylimide anion
Molecular Weight (g/mol): 444.41
Molecular Formula: C13H18F6N2O4S2
InChi Key: IGABCHQKFATAOI-UHFFFAOYSA-N
SMILES: CC[N+](C)(C)CC1=CC=CC=C1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results