accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNOHNOClF

CAS RN 1186372-20-2

IUPAC Name: 1-(2-chloro-6-fluorobenzoyl)-3-(9H-fluoren-9-yl)urea
Synonyms: 1-(2-chloro-6-fluorobenzoyl)-3-(9H-fluoren-9-yl)urea
Molecular Weight (g/mol): 380.8
Molecular Formula: C21H14ClFN2O2
InChi Key: SFNLLCUAISZNRV-UHFFFAOYSA-N
SMILES: FC1=C(C(=O)NC(=O)NC2C3=CC=CC=C3C3=CC=CC=C23)C(Cl)=CC=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results