Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Cyclophilin A Inhibitor

Catalog No. 23983610MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-983-610MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-983-610MG Supplier MilliporeSigma™ Supplier No. 23983610MG
Add to Cart
Add to Cart

The Cyclophilin A Inhibitor, also referenced under CAS 1186372-20-2, controls the biological activity of Cyclophilin A. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.

Acts as an inhibitor of cyclophilin A (IC50 = 1.5nM), which is approximately 27 times more potent than that of Cyclosporin A, in in vitro assays.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 95% by HPLC
CAS 1186372-20-2
Molecular Formula C21H14ClFN2O2
Formula Weight 380.8g/mol
Solubility DMSO
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.