CAS RN 1186648-22-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ MK-2 Inhibitor III
The MK-2 Inhibitor III, also referenced under CAS 1186648-22-5, controls the biological activity of MK-2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.