accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOCH3OOCH3

CAS RN 1187-74-2

IUPAC Name: 1,4-diethyl 2-(2-oxopropyl)butanedioate
Synonyms: 1,4-diethyl 2-(2-oxopropyl)butanedioate
Molecular Weight (g/mol): 230.26
Molecular Formula: C11H18O5
InChi Key: AAPVOVKOHNCXJA-UHFFFAOYNA-N
SMILES: CCOC(=O)CC(CC(C)=O)C(=O)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)