CAS RN 1187763-68-3
IUPAC Name:
2,6-dibromo-9,10-bis(4-tert-butylphenyl)anthracene
Synonyms:
2,6-dibromo-9,10-bis(4-tert-butylphenyl)anthracene
Molecular Weight (g/mol):
600.44
Molecular Formula:
C34H32Br2
InChi Key:
PVNXKVGTMVCYLK-UHFFFAOYSA-N
SMILES:
CC(C)(C)C1=CC=C(C=C1)C1=C2C=CC(Br)=CC2=C(C2=CC=C(C=C2)C(C)(C)C)C2=CC=C(Br)C=C12
1
–
1
of
1
results
1
2,6-Dibromo-9,10-bis(4-tert-butylphenyl)anthracene 99.0+%, TCI America™
CAS: 1187763-68-3 Molecular Formula: C34H32Br2 Molecular Weight (g/mol): 600.438 InChI Key: PVNXKVGTMVCYLK-UHFFFAOYSA-N PubChem CID: 118988970 IUPAC Name: 2,6-dibromo-9,10-bis(4-tert-butylphenyl)anthracene SMILES: CC(C)(C)C1=CC=C(C=C1)C2=C3C=C(C=CC3=C(C4=C2C=CC(=C4)Br)C5=CC=C(C=C5)C(C)(C)C)Br