accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1188423-16-6

IUPAC Name: 4,9,14,20,25,27,29,30,32,34-decamethoxyhexacyclo[21.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹]pentatriaconta-1(25),3,5,8,10,13,15,18,20,23,26,28,30,32,34-pentadecaene
Synonyms: 4,9,14,20,25,27,29,30,32,34-decamethoxyhexacyclo[21.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹]pentatriaconta-1(25),3,5,8,10,13,15,18,20,23,26,28,30,32,34-pentadecaene
Molecular Weight (g/mol): 750.89
Molecular Formula: C45H50O10
InChi Key: HJDHAGKELBPBLT-UHFFFAOYSA-N
SMILES: COC1=CC2=C(OC)C=C1CC1=CC(OC)=C(CC3=CC(OC)=C(CC4=C(OC)C=C(CC5=C(OC)C=C(C2)C(OC)=C5)C(OC)=C4)C=C3OC)C=C1OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results