accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1188990-02-4

IUPAC Name: {5-bromofuro[2,3-b]pyridin-2-yl}methanol
Synonyms: {5-bromofuro[2,3-b]pyridin-2-yl}methanol
Molecular Weight (g/mol): 228.05
Molecular Formula: C8H6BrNO2
InChi Key: SJASNTWQPIYYBY-UHFFFAOYSA-N
SMILES: OCC1=CC2=CC(Br)=CN=C2O1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results