accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONH(S)CH3H3CN(S)ON(S)O

CAS RN 118974-02-0

IUPAC Name: (1S,12S,15S)-7-methoxy-12-(2-methylprop-1-en-1-yl)-10,13,19-triazapentacyclo[11.7.0.0³,¹¹.0⁴,⁹.0¹⁵,¹⁹]icosa-3(11),4,6,8-tetraene-14,20-dione
Synonyms: (1S,12S,15S)-7-methoxy-12-(2-methylprop-1-en-1-yl)-10,13,19-triazapentacyclo[11.7.0.0³,¹¹.0⁴,⁹.0¹⁵,¹⁹]icosa-3(11),4,6,8-tetraene-14,20-dione
Molecular Weight (g/mol): 379.46
Molecular Formula: C22H25N3O3
InChi Key: DBEYVIGIPJSTOR-FHWLQOOXSA-N
SMILES: COC1=CC=C2C(NC3=C2C[C@@H]2N([C@H]3C=C(C)C)C(=O)[C@@H]3CCCN3C2=O)=C1
Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results