Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fumitremorgin C, Tocris Bioscience™

Catalog No. 2876250U
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-762-50U 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 28-762-50U Supplier R&D Systems Supplier No. 2876/250U
This item is not returnable. View return policy

Inhibitor of BCRP-mediated multidrug resistance

Chemosensitizing agent; selectively reverses BCRP-mediated multidrug resistance to mitoxantrone (Cat. No. 4250), doxorubicin (Cat. No. 2252) and topotecan (Cat. No. 4562) in transfected MCF-7 cells. Does not reverse resistance in P-gp- or MRP-positive cells. Analog ( Ko 143, Cat. No. 3241) also available.

Chemical Identifiers

CAS 118974-02-0
Molecular Formula C22H25N3O3
Molecular Weight (g/mol) 379.46
InChI Key DBEYVIGIPJSTOR-FHWLQOOXSA-N
Synonym fumitremorgin c, fumitremorgin c, aspergillus. fumigatus, fumitremorgin-c, sm-q, 5as,12s,14as-9-methoxy-12-2-methylprop-1-en-1-yl-1,2,3,5a,6,11,12,14a-octahydro-5h,14h-pyrrolo 1,2:4',5' pyrazino 1',2':1,6 pyrido 3,4-b indole-5,14-dione, 5h,14h-pyrrolo 1,2:4',5' pyrazino 1',2':1,6 pyrido 3,4-b indole-5,14-dione,1,2,3,5a,6,11,12,14a-octahydro-9-methoxy-12-2-methyl-1-propen-1-yl-, 5as,12s,14as, fumitremorgin c, from neosartorya fischeri hplc and tlc , film, 1s,12s,15s-7-methoxy-12-2-methylprop-1-en-1-yl-10,13,19-triazapentacyclo 11.7.0.0; 3,11 .0; 4,9 .0; 15,19 icosa-3 11 ,4,6,8-tetraene-14,20-dione, 5as,12s,14as-1,2,3,5a,6,11,12,14a-octahydro-9-methoxy-12-2-methyl-1-propen-1-yl-5h,14h-pyrrolo 1,2:4',5' pyrazino 1',2':1,6 pyrido 3,4-b indole-5,14-dione
PubChem CID 403923
ChEBI CHEBI:72763
SMILES CC(=CC1C2=C(CC3N1C(=O)C4CCCN4C3=O)C5=C(N2)C=C(C=C5)OC)C

Specifications

Quantity 250 μg
Formula Weight 379.45
Percent Purity >98%
Chemical Name or Material Fumitremorgin C
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.