CAS RN 119-64-2
1,2,3,4-Tetrahydronaphthalene, 97%
CAS: 119-64-2 Molecular Formula: C10H12 Molecular Weight (g/mol): 132.21 MDL Number: MFCD00001733 InChI Key: CXWXQJXEFPUFDZ-UHFFFAOYSA-N Synonym: tetralin,benzocyclohexane,tetrahydronaphthalene,bacticin,tetraline,tetranap,naphthalene, 1,2,3,4-tetrahydro,tetralina,naphthalene 1,2,3,4-tetrahydride,tetralene PubChem CID: 8404 ChEBI: CHEBI:35008 IUPAC Name: 1,2,3,4-tetrahydronaphthalene SMILES: C1CCC2=CC=CC=C2C1
1,2,3,4-Tetrahydronaphthalene, 98+%, pure
CAS: 119-64-2 Molecular Formula: C10H12 Molecular Weight (g/mol): 132.21 MDL Number: MFCD00001733 InChI Key: CXWXQJXEFPUFDZ-UHFFFAOYSA-N Synonym: tetralin,benzocyclohexane,tetrahydronaphthalene,bacticin,tetraline,tetranap,naphthalene, 1,2,3,4-tetrahydro,tetralina,naphthalene 1,2,3,4-tetrahydride,tetralene PubChem CID: 8404 ChEBI: CHEBI:35008 IUPAC Name: 1,2,3,4-tetrahydronaphthalene SMILES: C1CCC2=CC=CC=C2C1
1,2,3,4-Tetrahydronaphthalene 98.0+%, TCI America™
CAS: 119-64-2 Molecular Formula: C10H12 Molecular Weight (g/mol): 132.21 MDL Number: MFCD00001733 InChI Key: CXWXQJXEFPUFDZ-UHFFFAOYSA-N Synonym: tetralin,benzocyclohexane,tetrahydronaphthalene,bacticin,tetraline,tetranap,naphthalene, 1,2,3,4-tetrahydro,tetralina,naphthalene 1,2,3,4-tetrahydride,tetralene PubChem CID: 8404 ChEBI: CHEBI:35008 IUPAC Name: 1,2,3,4-tetrahydronaphthalene SMILES: C1CCC2=CC=CC=C2C1
1,2,3,4-Tetrahydronaphthalene 97.0+%, TCI America™
CAS: 119-64-2 Molecular Formula: C10H12 Molecular Weight (g/mol): 132.21 MDL Number: MFCD00001733 InChI Key: CXWXQJXEFPUFDZ-UHFFFAOYSA-N Synonym: tetralin,benzocyclohexane,tetrahydronaphthalene,bacticin,tetraline,tetranap,naphthalene, 1,2,3,4-tetrahydro,tetralina,naphthalene 1,2,3,4-tetrahydride,tetralene PubChem CID: 8404 ChEBI: CHEBI:35008 IUPAC Name: 1,2,3,4-tetrahydronaphthalene SMILES: C1CCC2=CC=CC=C2C1
1,2,3,4-Tetrahydronaphthalene (Laboratory), Fisher Chemical
CAS: 119-64-2 Molecular Formula: C10H12 Molecular Weight (g/mol): 132.21 MDL Number: MFCD00001733 InChI Key: CXWXQJXEFPUFDZ-UHFFFAOYSA-N Synonym: tetralin,benzocyclohexane,tetrahydronaphthalene,bacticin,tetraline,tetranap,naphthalene, 1,2,3,4-tetrahydro,tetralina,naphthalene 1,2,3,4-tetrahydride,tetralene PubChem CID: 8404 ChEBI: CHEBI:35008 IUPAC Name: 1,2,3,4-tetrahydronaphthalene SMILES: C1CCC2=CC=CC=C2C1
1,2,3,4-Tetrahydronaphthalene, 98+%, Thermo Scientific™
CAS: 119-64-2 Molecular Formula: C10H12 Molecular Weight (g/mol): 132.21 MDL Number: MFCD00001733 InChI Key: CXWXQJXEFPUFDZ-UHFFFAOYSA-N Synonym: tetralin,benzocyclohexane,tetrahydronaphthalene,bacticin,tetraline,tetranap,naphthalene, 1,2,3,4-tetrahydro,tetralina,naphthalene 1,2,3,4-tetrahydride,tetralene PubChem CID: 8404 ChEBI: CHEBI:35008 IUPAC Name: 1,2,3,4-tetrahydronaphthalene SMILES: C1CCC2=CC=CC=C2C1
1,2,3,4-Tetrahydronaphthalene in Dimethyl Sulfoxide, SPEX CertiPrep™
CAS: 119-64-2 Molecular Formula: C10H12 Molecular Weight (g/mol): 132.21 MDL Number: MFCD00001733 InChI Key: CXWXQJXEFPUFDZ-UHFFFAOYSA-N PubChem CID: 8404 ChEBI: CHEBI:35008 IUPAC Name: 1,2,3,4-tetrahydronaphthalene SMILES: C1CCC2=CC=CC=C2C1
1,2,3,4-Tetrahydronaphthalene, SPEX CertiPrep™
CAS: 119-64-2 Molecular Formula: C10H12 Molecular Weight (g/mol): 132.21 MDL Number: MFCD00001733 InChI Key: CXWXQJXEFPUFDZ-UHFFFAOYSA-N PubChem CID: 8404 ChEBI: CHEBI:35008 IUPAC Name: 1,2,3,4-tetrahydronaphthalene SMILES: C1CCC2=CC=CC=C2C1