Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,4-Tetrahydronaphthalene, 98+%, Thermo Scientific™

Catalog No. AC146730000
Catalog No. AC146730000 Supplier Thermo Scientific Chemicals Supplier No. 146730000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-64-2
Molecular Formula C10H12
Molecular Weight (g/mol) 132.21
MDL Number MFCD00001733
InChI Key CXWXQJXEFPUFDZ-UHFFFAOYSA-N
Synonym tetralin, benzocyclohexane, tetrahydronaphthalene, bacticin, tetraline, tetranap, naphthalene, 1,2,3,4-tetrahydro, tetralina, naphthalene 1,2,3,4-tetrahydride, tetralene
PubChem CID 8404
ChEBI CHEBI:35008
IUPAC Name 1,2,3,4-tetrahydronaphthalene
SMILES C1CCC2=CC=CC=C2C1

Specifications

Melting Point -35.0°C
Density 0.9730g/mL
Boiling Point 207.0°C
Flash Point 77°C
Refractive Index 1.5400 to 1.5420
Linear Formula C10H12
Specific Gravity 0.973
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with most common organic solvents
Formula Weight 132.2
Grade Pure
Chemical Name or Material 1,2,3,4-Tetrahydronaphthalene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.