accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOHN

CAS RN 119-75-5

IUPAC Name: 2-nitro-N-phenylaniline
Synonyms: 2-nitrodiphenylamine
Molecular Weight (g/mol): 214.22
Molecular Formula: C12H10N2O2
InChi Key: RUKISNQKOIKZGT-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results