Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrodiphenylamine, MP Biomedicals

Catalog No. ICN210318 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN210318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN210318 Supplier MP Biomedicals, Inc Supplier No. 0521031850
Add to Cart
Add to Cart

Chemical Identifiers

CAS 119-75-5
Molecular Formula C12H10N2O2
Molecular Weight (g/mol) 214.224
InChI Key RUKISNQKOIKZGT-UHFFFAOYSA-N
Synonym 2-nitrodiphenylamine, o-nitrodiphenylamine, nitrodiphenylamine, benzenamine, 2-nitro-n-phenyl, sudan yellow 1339, n-phenyl-o-nitroaniline, o-nitro-n-phenylaniline, diphenylamine, 2-nitro, 2-nitrophenyl phenylamine, 2-nitrodophenylamine
PubChem CID 8407
IUPAC Name 2-nitro-N-phenylaniline
SMILES C1=CC=C(C=C1)NC2=CC=CC=C2[N+](=O)[O-]

Specifications

Quantity 50 g
Formula Weight 214.22
Chemical Name or Material 2-Nitrodiphenylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.