accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1190-14-3

IUPAC Name: (4-bromobutyl)phosphonic acid
Synonyms: 4-bromobutylphosphonic acid
Molecular Weight (g/mol): 217
Molecular Formula: C4H10BrO3P
InChi Key: FXFARGAYPHUQQL-UHFFFAOYSA-N
SMILES: OP(O)(=O)CCCCBr
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results