Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Bromobutyl)phosphonic Acid 98.0+%, TCI America™
SDP

Catalog No. B5014200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5014200MG 200 mg
B50141G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5014200MG Supplier TCI America Supplier No. B5014200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1190-14-3
Molecular Formula C4H10BrO3P
Molecular Weight (g/mol) 217.00
MDL Number MFCD00466589
InChI Key FXFARGAYPHUQQL-UHFFFAOYSA-N
PubChem CID 18409311
IUPAC Name (4-bromobutyl)phosphonic acid
SMILES OP(O)(=O)CCCCBr

Specifications

Melting Point 129°C
Color White
Quantity 200 mg
UN Number 3261
Formula Weight 217.00
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (4-Bromobutyl)phosphonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.