CAS RN 1191063-57-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Carboxy-5-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
CAS: 1191063-57-6 Synonym: 2-Carboxy-5-fluorophenylboronic acid pinacol ester