accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1192-53-6

IUPAC Name: 4,5-dichloro-1-methyl-1H-imidazole
Synonyms: 4,5-dichloro-1-methylimidazole
Molecular Weight (g/mol): 150.99
Molecular Formula: C4H4Cl2N2
InChi Key: VVYQLPWYFQBZOL-UHFFFAOYSA-N
SMILES: CN1C=NC(Cl)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results