CAS RN 1192620-83-9
CAS RN 1192620-83-9
IUPAC Name:
chlororutheniumylium; {1,2-diphenyl-2-[(3-phenylpropyl)amino]ethyl}(4-methylbenzenesulfonyl)azanide
Synonyms:
chlororutheniumylium; {1,2-diphenyl-2-[(3-phenylpropyl)amino]ethyl}(4-methylbenzenesulfonyl)azanide
Molecular Weight (g/mol):
620.17
Molecular Formula:
C30H31ClN2O2RuS
InChi Key:
MDABGVLQRDDWLY-UHFFFAOYNA-M
SMILES:
Cl[Ru+].CC1=CC=C(C=C1)S(=O)(=O)[N-]C(C(NCCCC1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
Sigma Aldrich [(R,R)-Teth-Tsdpen Rucl]
Sigma Aldrich [(R,R)-Teth-Tsdpen Rucl]
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.