accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHONHN

CAS RN 1193-24-4

IUPAC Name: 6-hydroxy-3,4-dihydropyrimidin-4-one
Synonyms: 6-hydroxy-3H-pyrimidin-4-one
Molecular Weight (g/mol): 112.09
Molecular Formula: C4H4N2O2
InChi Key: DUFGYCAXVIUXIP-UHFFFAOYSA-N
SMILES: OC1=CC(=O)NC=N1
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (6)
  • (2)
  • (7)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)