CAS RN 1193-82-4
Methyl phenyl sulfoxide, 98+%
CAS: 1193-82-4 Molecular Formula: C7H8OS Molecular Weight (g/mol): 140.2 MDL Number: MFCD00002088 InChI Key: JXTGICXCHWMCPM-UHFFFAOYSA-N Synonym: methyl phenyl sulfoxide,methylsulfinyl benzene,thioanisole s-oxide,phenyl methyl sulfoxide,sulfoxide, methyl phenyl,benzene, methylsulfinyl,methanesulfinylbenzene,methylphenylsulfoxide,methylsulphinyl benzene PubChem CID: 14516 IUPAC Name: methylsulfinylbenzene SMILES: CS(=O)C1=CC=CC=C1
Methyl phenyl sulfoxide, 98+%
CAS: 1193-82-4 Molecular Formula: C7H8OS Molecular Weight (g/mol): 140.21 MDL Number: MFCD00002088 InChI Key: JXTGICXCHWMCPM-UHFFFAOYSA-N Synonym: methyl phenyl sulfoxide,methylsulfinyl benzene,thioanisole s-oxide,phenyl methyl sulfoxide,sulfoxide, methyl phenyl,benzene, methylsulfinyl,methanesulfinylbenzene,methylphenylsulfoxide,methylsulphinyl benzene PubChem CID: 14516 IUPAC Name: methylsulfinylbenzene SMILES: CS(=O)C1=CC=CC=C1
Methyl Phenyl Sulfoxide 98.0+%, TCI America™
CAS: 1193-82-4 Molecular Formula: C7H8OS Molecular Weight (g/mol): 140.2 MDL Number: MFCD00002088 InChI Key: JXTGICXCHWMCPM-UHFFFAOYSA-N Synonym: methyl phenyl sulfoxide,methylsulfinyl benzene,thioanisole s-oxide,phenyl methyl sulfoxide,sulfoxide, methyl phenyl,benzene, methylsulfinyl,methanesulfinylbenzene,methylphenylsulfoxide,methylsulphinyl benzene PubChem CID: 14516 IUPAC Name: methylsulfinylbenzene SMILES: CS(=O)C1=CC=CC=C1
Sigma Aldrich Methyl phenyl sulfoxide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
MP Biomedicals, Inc Methyl Phenyl Sulfoxide, MP Biomedicals
CAS: 1193-82-4 Molecular Formula: C7H8OS Molecular Weight (g/mol): 140.2 InChI Key: JXTGICXCHWMCPM-UHFFFAOYSA-N Synonym: methyl phenyl sulfoxide,methylsulfinyl benzene,thioanisole s-oxide,phenyl methyl sulfoxide,sulfoxide, methyl phenyl,benzene, methylsulfinyl,methanesulfinylbenzene,methylphenylsulfoxide,methylsulphinyl benzene PubChem CID: 14516 IUPAC Name: methylsulfinylbenzene SMILES: CS(=O)C1=CC=CC=C1