Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Phenyl Sulfoxide, MP Biomedicals

Catalog No. ICN228158 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN228158 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN228158 Supplier MP Biomedicals, Inc Supplier No. 0522815805

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1193-82-4
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.2
InChI Key JXTGICXCHWMCPM-UHFFFAOYSA-N
Synonym methyl phenyl sulfoxide, methylsulfinyl benzene, thioanisole s-oxide, phenyl methyl sulfoxide, sulfoxide, methyl phenyl, benzene, methylsulfinyl, methanesulfinylbenzene, methylphenylsulfoxide, methylsulphinyl benzene
PubChem CID 14516
IUPAC Name methylsulfinylbenzene
SMILES CS(=O)C1=CC=CC=C1

Specifications

Quantity 10 g
Formula Weight 140.2
Chemical Name or Material Methyl Phenyl Sulfoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.