accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNOH(E/Z)

CAS RN 1193-96-0

IUPAC Name: N-[(pyridin-2-yl)methylidene]hydroxylamine
Synonyms: 2-pyridinealdoxime
Molecular Weight (g/mol): 122.13
Molecular Formula: C6H6N2O
InChi Key: MTFJSAGADRTKCI-UHFFFAOYSA-N
SMILES: ON=CC1=CC=CC=N1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)