Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich syn-2-Pyridinealdoxime

Catalog No. P5820025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P5820025G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P5820025G Supplier Sigma Aldrich Supplier No. P5820025G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

9-Decen-1-ol undergoes oxidation to give 9-decenoic in aqueous media (dioxane-water mixtures) in the presence of supported palladium or platinum catalysts.9-Decen-1-ol was used in the preparation of semifluorinated acids required for the synthesis of poly(styrene-b-semifluorinated isoprene) block copolymers with -CF2H-terminated side groups.

Specifications

CAS 1193-96-0
Melting Point 110°C to 112°C (lit.)
Molecular Formula C6H6N2O
Linear Formula C6H6N2O
MDL Number MFCD00006354
Quantity 10 g
Molecular Weight (g/mol) 122.12
Percent Purity ≥99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.