accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1193447-34-5

IUPAC Name: 6-{2-[4-(dimethylamino)phenyl]ethenyl}-4-{2-[4-(dimethyliminiumyl)cyclohexa-2,5-dien-1-ylidene]ethylidene}-2,2-difluoro-2,4-dihydro-1,3,2-dioxaborinin-2-uide
Synonyms: 4-{2-[4-(dimethylamino)phenyl]ethenyl}-6-{2-[4-(dimethyliminio)cyclohexa-2,5-dien-1-ylidene]ethylidene}-2,2-difluoro-2H-1,3,2-dioxaborinin-2-uide
Molecular Weight (g/mol): 410.27
Molecular Formula: C23H25BF2N2O2
InChi Key: PVXQJYXODFZTBR-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=CC2=CC(O[B-](F)(F)O2)=CC=C2C=CC(C=C2)=[N+](C)C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results