accessibility menu, dialog, popup

UserName

Viewing profile as user:
HO(R)NH(R)OH(S)OHHO(R)HO

CAS RN 119557-99-2

IUPAC Name: (2R,3R,5S,6R)-2,6-bis(hydroxymethyl)piperidine-3,4,5-triol
Synonyms: (2R,3R,5S,6R)-2,6-bis(hydroxymethyl)piperidine-3,4,5-triol
Molecular Weight (g/mol): 193.2
Molecular Formula: C7H15NO5
InChi Key: CLVUFWXGNIFGNC-QTSLKERKSA-N
SMILES: OC[C@H]1N[C@H](CO)[C@H](O)C(O)[C@@H]1O
Color 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results