Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Homonojirimycin 98.0+%, TCI America™
SDP

Catalog No. H114410MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H114410MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H114410MG Supplier TCI America Supplier No. H114410MG

Chemical Identifiers

CAS 119557-99-2
Molecular Formula C7H15NO5
Molecular Weight (g/mol) 193.20
MDL Number MFCD06797112
InChI Key CLVUFWXGNIFGNC-QTSLKERKSA-N
PubChem CID 159496
IUPAC Name (2R,3R,5S,6R)-2,6-bis(hydroxymethyl)piperidine-3,4,5-triol
SMILES OC[C@H]1N[C@H](CO)[C@H](O)C(O)[C@@H]1O

Specifications

Type alpha-Homonojirimycin
Physical Form Crystalline Powder
Melting Point 207°C
Color White
Quantity 10 mg
Formula Weight 193.20
Percent Purity ≥98.0% (HPLC)
Chemical Name or Material alpha-Homonojirimycin
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.