CAS RN 1196-58-3
IUPAC Name:
(pentan-3-yl)benzene
Synonyms:
benzene, (1-ethylpropyl)-
Molecular Weight (g/mol):
148.25
Molecular Formula:
C11H16
InChi Key:
PBWHJRFXUPLZDS-UHFFFAOYSA-N
SMILES:
CCC(CC)C1=CC=CC=C1
1
–
1
of
1
results
1
(1-Ethylpropyl)benzene 99.0+%, TCI America™
CAS: 1196-58-3 Molecular Formula: C11H16 Molecular Weight (g/mol): 148.25 MDL Number: MFCD00048663 InChI Key: PBWHJRFXUPLZDS-UHFFFAOYSA-N Synonym: 3-Phenylpentane PubChem CID: 14527 IUPAC Name: (pentan-3-yl)benzene SMILES: CCC(CC)C1=CC=CC=C1