Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1-Ethylpropyl)benzene 99.0+%, TCI America™
SDP

Catalog No. E03051ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E03051ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E03051ML Supplier TCI America Supplier No. E03051ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1196-58-3
Molecular Formula C11H16
Molecular Weight (g/mol) 148.25
MDL Number MFCD00048663
InChI Key PBWHJRFXUPLZDS-UHFFFAOYSA-N
Synonym 3-Phenylpentane
PubChem CID 14527
IUPAC Name (pentan-3-yl)benzene
SMILES CCC(CC)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 191°C
Quantity 1 mL
UN Number 3295
Formula Weight 148.25
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material (1-Ethylpropyl)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.