accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNCH3

CAS RN 1196-79-8

IUPAC Name: 2,5-dimethyl-1H-indole
Synonyms: 1H-indole, 2,5-dimethyl-
Molecular Weight (g/mol): 145.21
Molecular Formula: C10H11N
InChi Key: ZFLFWZRPMDXJCW-UHFFFAOYSA-N
SMILES: CC1=CC2=CC(C)=CC=C2N1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)