Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylindole 97.0+%, TCI America™
SDP

Catalog No. D36985G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36985G 5 g
D36981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36985G Supplier TCI America Supplier No. D36985G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1196-79-8
Molecular Formula C10H11N
Molecular Weight (g/mol) 145.205
MDL Number MFCD00005621
InChI Key ZFLFWZRPMDXJCW-UHFFFAOYSA-N
Synonym 2,5-dimethylindole, 1h-indole, 2,5-dimethyl, indole, 2,5-dimethyl, unii-etu94v283m, 2,5-dimethyl indole, pubchem7330, acmc-1bvpm
PubChem CID 70965
IUPAC Name 2,5-dimethyl-1H-indole
SMILES CC1=CC2=C(C=C1)NC(=C2)C

Specifications

Melting Point 115°C
Quantity 5 g
Formula Weight 145.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dimethylindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.