accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1197-22-4

IUPAC Name: N-phenylmethanesulfonamide
Synonyms: N-phenyl methanesulfonamide
Molecular Weight (g/mol): 171.21
Molecular Formula: C7H9NO2S
InChi Key: LBTPIFQNEKOAIM-UHFFFAOYSA-N
SMILES: CS(=O)(=O)NC1=CC=CC=C1
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)