Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Phenylmethanesulfonamide, 98%, Thermo Scientific™

Catalog No. AC417640025
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC417640025 2.5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC417640025 Supplier Thermo Scientific Chemicals Supplier No. 417640025
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1197-22-4
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.21
MDL Number MFCD00043782
InChI Key LBTPIFQNEKOAIM-UHFFFAOYSA-N
Synonym methanesulfonamide, n-phenyl, methanesulfonanilide, methanesulfanilide, mesylaniline, n-mesylaniline, n-methylsulphonylaniline, n-phenyl-methanesulfonamide, methylsulfonyl phenylamine, mesylanilide, methylsulfonyl aniline
PubChem CID 70970
IUPAC Name N-phenylmethanesulfonamide
SMILES CS(=O)(=O)NC1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 93°C to 97°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3SO2NHC6H5
Quantity 2.5 g
Formula Weight 171.22
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material N-Phenylmethanesulfonamide, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.