CAS RN 1198-63-6
IUPAC Name:
tetrafluorobenzene-1,3-diamine
Synonyms:
tetrafluorobenzene-1,3-diamine
Molecular Weight (g/mol):
180.11
Molecular Formula:
C6H4F4N2
InChi Key:
FXGQUGCFZKMIJW-UHFFFAOYSA-N
SMILES:
NC1=C(F)C(F)=C(F)C(N)=C1F
1
–
1
of
1
results
1
2,4,5,6-Tetrafluoro-1,3-phenylenediamine 95.0+%, TCI America™
CAS: 1198-63-6 Molecular Formula: C6H4F4N2 Molecular Weight (g/mol): 180.11 MDL Number: MFCD00039764 InChI Key: FXGQUGCFZKMIJW-UHFFFAOYSA-N Synonym: 1,3-Diamino-2,4,5,6-tetrafluorobenzene PubChem CID: 70978 IUPAC Name: tetrafluorobenzene-1,3-diamine SMILES: NC1=C(F)C(F)=C(F)C(N)=C1F