accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 119818-40-5

IUPAC Name: ethyl docosa-7,10,13,16,19-pentaenoate
Synonyms: ethyl docosa-7,10,13,16,19-pentaenoate
Molecular Weight (g/mol): 358.57
Molecular Formula: C24H38O2
InChi Key: VCSQUSNNIFZJAP-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCCCC=CCC=CCC=CCC=CCC=CCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results