Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl all cis-7,10,13,16,19-Docosapentaenoate 96.0+%, TCI America™
SDP

Catalog No. D2964100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2964100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D2964100MG Supplier TCI America Supplier No. D2964100MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 119818-40-5
Molecular Formula C24H38O2
Molecular Weight (g/mol) 358.57
MDL Number MFCD02259225
InChI Key VCSQUSNNIFZJAP-UHFFFAOYSA-N
Synonym all cis-7,10,13,16,19-Docosapentaenoic Acid Ethyl Ester, DPA Ethyl Ester
PubChem CID 12054543
ChEBI CHEBI:84937
IUPAC Name ethyl docosa-7,10,13,16,19-pentaenoate
SMILES CCOC(=O)CCCCCC=CCC=CCC=CCC=CCC=CCC

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 358.57
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl all cis-7,10,13,16,19-Docosapentaenoate
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.