accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3O

CAS RN 1198467-57-0

IUPAC Name: 2,6-bis(dodecyloxy)benzaldehyde
Synonyms: 2,6-bis(dodecyloxy)benzaldehyde
Molecular Weight (g/mol): 474.77
Molecular Formula: C31H54O3
InChi Key: QPAKGKFSBBAHMS-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCOC1=CC=CC(OCCCCCCCCCCCC)=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results