Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Bis(dodecyloxy)benzaldehyde, 97%, Thermo Scientific Chemicals

Catalog No. AAH64087MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64087MD 250 mg
AAH6408703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH64087MD Supplier Thermo Scientific Chemicals Supplier No. H64087MD

missing translation for 'validMainframeMsg'

CAS: 1198467-57-0 | Molecular Formula: C31H54O3 | Molecular Weight: 474.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1198467-57-0
Molecular Formula C31H54O3
Molecular Weight (g/mol) 474.77
MDL Number MFCD25372091
InChI Key QPAKGKFSBBAHMS-UHFFFAOYSA-N
Synonym 2,6-bis dodecyloxy benzaldehyde, 2,6-di dodecyloxy benzaldehyde
PubChem CID 75201262
IUPAC Name 2,6-didodecoxybenzaldehyde
SMILES CCCCCCCCCCCCOC1=C(C(=CC=C1)OCCCCCCCCCCCC)C=O

Specifications

Color Yellow
Quantity 250 mg
Beilstein 20848961
Formula Weight 474.77
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2,6-Bis(dodecyloxy)benzaldehyde
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.