CAS RN 1199215-88-7
CAS RN 1199215-88-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Sigma Aldrich 3-Chloro-5-(trifluoromethyl)-1H-1,2,4-triazole
Sigma Aldrich 3-Chloro-5-(trifluoromethyl)-1H-1,2,4-triazole
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.