accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNN

CAS RN 120-03-6

IUPAC Name: 2-(4-methylphenyl)-1H-1,3-benzodiazole
Synonyms: 2-(4-methylphenyl)-1H-1,3-benzodiazole
Molecular Weight (g/mol): 208.26
Molecular Formula: C14H12N2
InChi Key: YJSWQFYMPODDTQ-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1=NC2=CC=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results