Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Methylphenyl)benzimidazole, 97%, Thermo Scientific™

Catalog No. AAH5889409
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5889409 10 g
AAH5889404 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5889409 Supplier Thermo Scientific Chemicals Supplier No. H5889409

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-03-6
Molecular Formula C14H12N2
Molecular Weight (g/mol) 208.264
MDL Number MFCD00209485
InChI Key YJSWQFYMPODDTQ-UHFFFAOYSA-N
Synonym 2-p-tolyl-1h-benzoimidazole, 2-4-methylphenyl-1h-benzimidazole, 2-4-methylphenyl benzimidazole, 2-p-tolyl-1h-benzo d imidazole, 1h-benzimidazole,2-4-methylphenyl, 2-4-methylphenyl-1h-1,3-benzodiazole, 1h-benzimidazole, 2-4-methylphenyl, 2-ptolyl-1h-benzoimidazole, 2-4-methylphenyl-1h-benzo d imidazole
PubChem CID 725982
IUPAC Name 2-(4-methylphenyl)-1H-benzimidazole
SMILES CC1=CC=C(C=C1)C2=NC3=CC=CC=C3N2

Specifications

Melting Point 265°C to 269°C
Color Brown
Quantity 10 g
Formula Weight 208.26
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-(4-Methylphenyl)benzimidazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.