accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3

CAS RN 120-14-9

IUPAC Name: 3,4-dimethoxybenzaldehyde
Synonyms: veratraldehyde protocatechuic aldehyde dimethyl ether protocatechualdehyde dimethyl ether veratrum aldehyde protocatechuecaldehyde dimethyl ether
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: WJUFSDZVCOTFON-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=O)C=C1OC
Boiling Point 
  • (10)
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (11)
Quantity 
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
Percent Purity 
  • (3)
  • (6)
  • (4)
Physical Form 
  • (3)
  • (4)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (1)
  • (3)
  • (1)
  • (3)
  • (7)
  • (10)
  • (1)
  • (3)
  • (3)
  • (11)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)
  • (6)
  • (4)
  • (3)
  • (4)
  • (1)
  • (1)