Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethoxybenzaldehyde, Spectrum™ Chemical
SDP

Catalog No. 18620639 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-639 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-620-639 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D2440100GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synonym: Veratraldehyde, Veratric Aldehyde, Methylvanillin

3,4-Dimethoxybenzaldehyde, also known as Methylvanillin or Veratric aldehyde, is an organic compound structurally related to benzaldehyde. It is used as an odorant and flavorant. Ungraded products supplied by Spectrum™ are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Chemical Identifiers

CAS 120-14-9
Molecular Weight (g/mol) 166.18

Specifications

Boiling Point 281°C (537.8°F)
Quantity 100 g
Grade Ungraded
Physical Form Powder or Waxy Solid
Chemical Name or Material 3,4-Dimethoxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.