accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 120-52-5

IUPAC Name: [(1E,4E)-4-[(benzoyloxy)imino]cyclohexa-2,5-dien-1-ylidene]amino benzoate
Synonyms: [(1E,4E)-4-[(benzoyloxy)imino]cyclohexa-2,5-dien-1-ylidene]amino benzoate
Molecular Weight (g/mol): 346.34
Molecular Formula: C20H14N2O4
InChi Key: WMVSVUVZSYRWIY-QGFZOGOGSA-N
SMILES: O=C(ON=C1C=CC(C=C1)=NOC(=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results