accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 120-53-6

IUPAC Name: 6-ethoxy-2,3-dihydro-1,3-benzothiazole-2-thione
Synonyms: 6-ethoxy-3H-1,3-benzothiazole-2-thione
Molecular Weight (g/mol): 211.3
Molecular Formula: C9H9NOS2
InChi Key: HOASVNMVYBSLSU-UHFFFAOYSA-N
SMILES: CCOC1=CC=C2NC(=S)SC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results