accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 120-55-8

IUPAC Name: 2-[2-(benzoyloxy)ethoxy]ethyl benzoate
Synonyms: diethylene glycol dibenzoate
Molecular Weight (g/mol): 314.34
Molecular Formula: C18H18O5
InChi Key: NXQMCAOPTPLPRL-UHFFFAOYSA-N
SMILES: O=C(OCCOCCOC(=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results