accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CHO

CAS RN 120-65-0

IUPAC Name: 2-[(dimethylamino)methyl]phenol
Synonyms: 2-[(dimethylamino)methyl]phenol
Molecular Weight (g/mol): 151.21
Molecular Formula: C9H13NO
InChi Key: FUIQBJHUESBZNU-UHFFFAOYSA-N
SMILES: CN(C)CC1=CC=CC=C1O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results