accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OHH3CCH3CH3

CAS RN 120-95-6

IUPAC Name: 2,4-bis(2-methylbutan-2-yl)phenol
Synonyms: 2,4-di-t-pentylphenol
Molecular Weight (g/mol): 234.38
Molecular Formula: C16H26O
InChi Key: WMVJWKURWRGJCI-UHFFFAOYSA-N
SMILES: CCC(C)(C)C1=CC=C(O)C(=C1)C(C)(C)CC
Percent Purity 
  • (4)
Physical Form 
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results